Assay Details
| Gene Symbol: Fgf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512038.2 | XP_003512086.1 | 2 - 3 | 813 | 87 | |
| XM_007652530.1 | XP_007650720.1 | 2 - 3 | 809 | 87 | ||
| GenBank mRNA | JI894807.1 | - | 2 - 3 | 796 | 87 | |
| JP041466.1 | - | 2 - 3 | 796 | 87 | ||
More Information
Set Membership: |
|

Fgf7
Genomic Map