Assay Details
| Gene Symbol: LOC100760352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513189.2 | XP_003513237.1 | 4 - 5 | 523 | 75 | |
| XM_003513190.2 | XP_003513238.1 | 4 - 5 | 493 | 75 | ||
| XM_007653504.1 | XP_007651694.1 | 5 - 6 | 553 | 75 | ||
| XM_007653505.1 | XP_007651695.1 | 4 - 5 | 491 | 75 | ||
| XM_007653506.1 | XP_007651696.1 | 4 - 5 | 487 | 75 | ||
| GenBank mRNA | JI877799.1 | - | 4 - 5 | 522 | 75 | |
| JP055504.1 | - | 4 - 5 | 522 | 75 | ||
More Information
Set Membership: |
|

LOC100760352
Genomic Map