Assay Details
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654308.1 | XP_007652498.1 | 7 - 8 | 2019 | 61 | |
XM_007654309.1 | XP_007652499.1 | 7 - 8 | 2017 | 61 | ||
XM_007654310.1 | XP_007652500.1 | 6 - 7 | 1891 | 61 | ||
XM_007654311.1 | XP_007652501.1 | 7 - 8 | 1920 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
