Assay Details
| Gene Symbol: Il18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007629383.1 | XP_007627573.1 | 5 - 6 | 454 | 97 | |
| XM_007629389.1 | XP_007627579.1 | 6 - 7 | 576 | 97 | ||
| XM_007629398.1 | XP_007627588.1 | 6 - 7 | 566 | 97 | ||
| XM_007629405.1 | XP_007627595.1 | 6 - 7 | 583 | 97 | ||
| XM_007629412.1 | XP_007627602.1 | 4 - 5 | 275 | 97 | ||
| GenBank mRNA | JI868559.1 | - | 6 - 7 | 708 | 97 | |
| JP061905.1 | - | 6 - 7 | 708 | 97 | ||
More Information
Set Membership: |
|

Il18
Genomic Map