Assay Details
| Gene Symbol: Cd59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497437.2 | XP_003497485.1 | 2 - 3 | 223 | 105 | |
| XM_007612154.1 | XP_007610344.1 | 2 - 3 | 485 | 105 | ||
| XM_007612163.1 | XP_007610353.1 | 2 - 3 | 225 | 105 | ||
| GenBank mRNA | JI871275.1 | - | 2 - 3 | 175 | 105 | |
More Information
Set Membership: |
|

Cd59
Genomic Map