Assay Details
| Gene Symbol: Atpif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498382.2 | XP_003498430.1 | 1 - 2 | 236 | 70 | |
| GenBank mRNA | JI894091.1 | - | 1 - 2 | 183 | 70 | |
| JP051073.1 | - | 1 - 2 | 183 | 70 | ||
More Information
Set Membership: |
|

Atpif1
Genomic Map