Assay Details
| Gene Symbol: Atpif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498382.2 | XP_003498430.1 | 2 - 3 | 330 | 73 | |
| GenBank mRNA | JI894091.1 | - | 2 - 3 | 277 | 73 | |
| JP051073.1 | - | 2 - 3 | 277 | 73 | ||
More Information
Set Membership: |
|

Atpif1
Genomic Map