Assay Details
| Gene Symbol: Emp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496684.2 | XP_003496732.1 | 1 - 2 | 179 | 66 | |
| GenBank mRNA | JI873007.1 | - | 1 - 2 | 160 | 66 | |
| JP057656.1 | - | 1 - 2 | 160 | 66 | ||
More Information
Set Membership: |
|

Emp2
Genomic Map