Assay Details
Gene Symbol: Odf3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497916.2 | XP_003497964.1 | 2 - 3 | 190 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
