Assay Details
| Gene Symbol: Eps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500911.2 | XP_003500959.1 | 16 - 17 | 1717 | 62 | |
| XM_007641642.1 | XP_007639832.1 | 17 - 18 | 1830 | 62 | ||
| XM_007641643.1 | XP_007639833.1 | 7 - 8 | 866 | 62 | ||
| GenBank mRNA | JI873028.1 | - | 4 - 5 | 502 | 62 | |
| JP048364.1 | - | 4 - 5 | 498 | 62 | ||
More Information
Set Membership: |
|

Eps8
Genomic Map