Assay Details
| Gene Symbol: Ndufb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501889.2 | XP_003501937.1 | 1 - 2 | 203 | 77 | |
| GenBank mRNA | JI873565.1 | - | 1 - 2 | 202 | 77 | |
| JP041437.1 | - | 1 - 2 | 202 | 77 | ||
More Information
Set Membership: |
|

Ndufb7
Genomic Map