Assay Details
| Gene Symbol: Sap30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504319.2 | XP_003504367.1 | 3 - 4 | 844 | 80 | |
| GenBank mRNA | JI867149.1 | - | 3 - 4 | 329 | 80 | |
| JP052987.1 | - | 3 - 4 | 329 | 80 | ||
More Information
Set Membership: |
|

Sap30
Genomic Map