Assay Details
| Gene Symbol: Rnls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505952.2 | XP_003506000.2 | 2 - 3 | 171 | 68 | |
| XM_007646437.1 | XP_007644627.1 | 2 - 3 | 286 | 68 | ||
| GenBank mRNA | JI880660.1 | - | 2 - 3 | 178 | 68 | |
More Information
Set Membership: |
|

Rnls
Genomic Map