Assay Details
| Gene Symbol: Suv420h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510377.2 | XP_003510425.1 | 7 - 8 | 913 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Suv420h1| Gene Symbol: Suv420h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510377.2 | XP_003510425.1 | 7 - 8 | 913 | 93 | |
Set Membership: |
|