Assay Details
| Gene Symbol: Timp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512242.2 | XP_003512290.1 | 4 - 5 | 423 | 58 | |
| GenBank mRNA | JI873443.1 | - | 4 - 5 | 434 | 58 | |
| JP061669.1 | - | 4 - 5 | 434 | 58 | ||
More Information
Set Membership: |
|

Timp1
Genomic Map