Assay Details
| Gene Symbol: LOC100754168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512578.2 | XP_003512626.1 | 4 - 5 | 527 | 103 | |
| GenBank mRNA | JI881472.1 | - | 3 - 4 | 246 | 103 | |
| JP053329.1 | - | 3 - 4 | 246 | 103 | ||
More Information
Set Membership: |
|

LOC100754168
Genomic Map