Assay Details
| Gene Symbol: LOC100754168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512578.2 | XP_003512626.1 | 6 - 7 | 834 | 69 | |
| GenBank mRNA | JI881474.1 | - | 1 - 2 | 140 | 69 | |
| JP053330.1 | - | 1 - 2 | 140 | 69 | ||
More Information
Set Membership: |
|

LOC100754168
Genomic Map