Assay Details
| Gene Symbol: LOC100764242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499888.2 | XP_003499936.1 | 5 - 6 | 689 | 87 | |
| XM_007640638.1 | XP_007638828.1 | 6 - 7 | 683 | 87 | ||
| XM_007640639.1 | XP_007638829.1 | 5 - 6 | 662 | 87 | ||
More Information
Set Membership: |
|

LOC100764242
Genomic Map