Assay Details
| Gene Symbol: LOC100764242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499888.2 | XP_003499936.1 | 7 - 8 | 953 | 56 | |
| XM_007640638.1 | XP_007638828.1 | 8 - 9 | 947 | 56 | ||
| XM_007640639.1 | XP_007638829.1 | 7 - 8 | 926 | 56 | ||
More Information
Set Membership: |
|

LOC100764242
Genomic Map