Assay Details
| Gene Symbol: Tsacc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502910.2 | XP_003502958.1 | 3 - 4 | 235 | 78 | |
| XM_007643527.1 | XP_007641717.1 | 3 - 4 | 519 | 78 | ||
| XM_007643528.1 | XP_007641718.1 | 3 - 4 | 519 | 78 | ||
| GenBank mRNA | JI882262.1 | - | 2 - 3 | 175 | 78 | |
| JP060517.1 | - | 2 - 3 | 175 | 78 | ||
More Information
Set Membership: |
|

Tsacc
Genomic Map