Assay Details
| Gene Symbol: Hn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642176.1 | XP_007640366.1 | 1 - 2 | 221 | 92 | |
| GenBank mRNA | JP043540.1 | - | 1 - 2 | 221 | 92 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hn1l| Gene Symbol: Hn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642176.1 | XP_007640366.1 | 1 - 2 | 221 | 92 | |
| GenBank mRNA | JP043540.1 | - | 1 - 2 | 221 | 92 | |