Assay Details
| Gene Symbol: Ppp1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640483.1 | XP_007638673.1 | 3 - 4 | 304 | 79 | |
| GenBank mRNA | JP051573.1 | - | 3 - 4 | 364 | 79 | |
More Information
Set Membership: |
|

Ppp1r2
Genomic Map