Assay Details
| Gene Symbol: Tctex1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495399.2 | XP_003495447.1 | 1 - 2 | 220 | 67 | |
| GenBank mRNA | JI870643.1 | - | 1 - 2 | 197 | 67 | |
More Information
Set Membership: |
|

Tctex1d2
Genomic Map