Assay Details
| Gene Symbol: Actr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498290.2 | XP_003498338.1 | 1 - 2 | 53 | 58 | |
| XM_007625677.1 | XP_007623867.1 | 1 - 2 | 53 | 58 | ||
More Information
Set Membership: |
|

Actr1b
Genomic Map