Assay Details
| Gene Symbol: Psmd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498443.2 | XP_003498491.1 | 2 - 3 | 337 | 61 | |
| GenBank mRNA | JI886423.1 | - | 2 - 3 | 282 | 61 | |
| JP045488.1 | - | 2 - 3 | 282 | 61 | ||
More Information
Set Membership: |
|

Psmd9
Genomic Map