Assay Details
| Gene Symbol: LOC100761917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498567.2 | XP_003498615.1 | 1 | 1418 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100761917| Gene Symbol: LOC100761917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498567.2 | XP_003498615.1 | 1 | 1418 | 86 | |
Set Membership: |
|