Assay Details
| Gene Symbol: Atp6v0b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502688.2 | XP_003502736.1 | 6 - 7 | 445 | 54 | |
| GenBank mRNA | JI875263.1 | - | 6 - 7 | 508 | 54 | |
More Information
Set Membership: |
|

Atp6v0b
Genomic Map