Assay Details
| Gene Symbol: LOC100763814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511505.1 | XP_003511553.1 | 2 - 3 | 485 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763814| Gene Symbol: LOC100763814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511505.1 | XP_003511553.1 | 2 - 3 | 485 | 59 | |
Set Membership: |
|