Assay Details
| Gene Symbol: Figf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643841.1 | XP_007642031.1 | 3 - 4 | 1264 | 102 | |
| XM_007643842.1 | XP_007642032.1 | 3 - 4 | 1264 | 102 | ||
| XM_007643843.1 | XP_007642033.1 | 3 - 4 | 427 | 102 | ||
| XM_007643844.1 | XP_007642034.1 | 3 - 4 | 1249 | 102 | ||
| GenBank mRNA | JI873754.1 | - | 3 - 4 | 594 | 102 | |
| JP043745.1 | - | 3 - 4 | 594 | 102 | ||
More Information
Set Membership: |
|

Figf
Genomic Map