Assay Details
| Gene Symbol: Tfdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641541.1 | XP_007639731.1 | 11 - 12 | 1197 | 76 | |
| XM_007641544.1 | XP_007639734.1 | 11 - 12 | 1185 | 76 | ||
| XM_007641548.1 | XP_007639738.1 | 10 - 11 | 1146 | 76 | ||
| XM_007641552.1 | XP_007639742.1 | 10 - 11 | 1122 | 76 | ||
| XM_007641558.1 | XP_007639748.1 | 9 - 10 | 985 | 76 | ||
| XM_007641564.1 | XP_007639754.1 | 9 - 10 | 995 | 76 | ||
| GenBank mRNA | JI880665.1 | - | 9 - 10 | 1200 | 76 | |
More Information
Set Membership: |
|

Tfdp2
Genomic Map