Assay Details
| Gene Symbol: Ntm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644438.1 | XP_007642628.1 | 4 - 5 | 659 | 67 | |
| XM_007644442.1 | XP_007642632.1 | 4 - 5 | 659 | 67 | ||
More Information
Set Membership: |
|

Ntm
Genomic Map