Assay Details
| Gene Symbol: LOC100772531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640544.1 | XP_007638734.1 | 10 - 11 | 1157 | 71 | |
| GenBank mRNA | JI880083.1 | - | 6 - 7 | 679 | 71 | |
| JP038201.1 | - | 7 - 8 | 786 | 71 | ||
More Information
Set Membership: |
|

LOC100772531
Genomic Map