Assay Details
| Gene Symbol: LOC100772531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640544.1 | XP_007638734.1 | 5 - 6 | 546 | 61 | |
| GenBank mRNA | JI880083.1 | - | 1 - 2 | 68 | 61 | |
| JP038201.1 | - | 2 - 3 | 175 | 61 | ||
More Information
Set Membership: |
|

LOC100772531
Genomic Map