Assay Details
| Gene Symbol: LOC100772531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640544.1 | XP_007638734.1 | 7 - 8 | 779 | 91 | |
| GenBank mRNA | JI880083.1 | - | 3 - 4 | 301 | 91 | |
| JP038201.1 | - | 4 - 5 | 408 | 91 | ||
More Information
Set Membership: |
|

LOC100772531
Genomic Map