Assay Details
| Gene Symbol: LOC100772531 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640544.1 | XP_007638734.1 | 9 - 10 | 992 | 63 | |
| GenBank mRNA | JI880083.1 | - | 5 - 6 | 514 | 63 | |
| JP038201.1 | - | 6 - 7 | 621 | 63 | ||
More Information
Set Membership: |
|

LOC100772531
Genomic Map