Assay Details
| Gene Symbol: Icam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642520.1 | XP_007640710.1 | 3 - 4 | 769 | 78 | |
| XM_007642521.1 | XP_007640711.1 | 4 - 5 | 704 | 78 | ||
More Information
Set Membership: |
|

Icam2
Genomic Map