Assay Details
| Gene Symbol: Arpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496855.2 | XP_003496903.1 | 5 - 6 | 596 | 64 | |
| XM_007653725.1 | XP_007651915.1 | 5 - 6 | 821 | 64 | ||
| GenBank mRNA | JI866089.1 | - | 5 - 6 | 899 | 64 | |
More Information
Set Membership: |
|

Arpc4
Genomic Map