Assay Details
| Gene Symbol: LOC100771037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509531.1 | XP_003509579.1 | 24 - 25 | 4105 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100771037| Gene Symbol: LOC100771037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509531.1 | XP_003509579.1 | 24 - 25 | 4105 | 110 | |
Set Membership: |
|