Assay Details
| Gene Symbol: LOC100758105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510138.2 | XP_003510186.1 | 2 - 3 | 461 | 78 | |
| XM_007650778.1 | XP_007648968.1 | 2 - 3 | 461 | 78 | ||
More Information
Set Membership: |
|

LOC100758105
Genomic Map