Assay Details
| Gene Symbol: LOC100751499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512386.2 | XP_003512434.1 | 5 - 6 | 808 | 87 | |
| GenBank mRNA | JI862118.1 | - | 3 - 4 | 477 | 87 | |
| JP048190.1 | - | 3 - 4 | 477 | 87 | ||
More Information
Set Membership: |
|

LOC100751499
Genomic Map