Assay Details
| Gene Symbol: LOC100768782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514724.2 | XP_003514772.1 | 1 - 2 | 309 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100768782| Gene Symbol: LOC100768782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514724.2 | XP_003514772.1 | 1 - 2 | 309 | 97 | |