Assay Details
| Gene Symbol: Arf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007646997.1 | XP_007645187.1 | 1 - 2 | 286 | 64 | |
| GenBank mRNA | JI862977.1 | - | 1 - 2 | 327 | 63 | |
| JP051204.1 | - | 1 - 2 | 328 | 64 | ||
More Information
Set Membership: |
|

Arf4
Genomic Map