Assay Details
| Gene Symbol: Tmigd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642447.1 | XP_007640637.1 | 4 - 5 | 629 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmigd2| Gene Symbol: Tmigd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642447.1 | XP_007640637.1 | 4 - 5 | 629 | 59 | |
Set Membership: |
|