Assay Details
| Gene Symbol: Cbx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650055.1 | XP_007648245.1 | 1 - 2 | 220 | 61 | |
| GenBank mRNA | JI875058.1 | - | 1 - 2 | 220 | 61 | |
More Information
Set Membership: |
|

Cbx6
Genomic Map