Assay Details
| Gene Symbol: Alg12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652917.1 | XP_007651107.1 | 11 - 12 | 1880 | 63 | |
| XM_007652918.1 | XP_007651108.1 | 11 - 12 | 1963 | 63 | ||
| XM_007652919.1 | XP_007651109.1 | 10 - 11 | 1660 | 63 | ||
| XM_007652920.1 | XP_007651110.1 | 10 - 11 | 1706 | 63 | ||
| GenBank mRNA | JI891076.1 | - | 3 - 4 | 470 | 63 | |
| JP063732.1 | - | 3 - 4 | 470 | 63 | ||
More Information
Set Membership: |
|

Alg12
Genomic Map