Assay Details
| Gene Symbol: Chmp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495079.2 | XP_003495127.1 | 4 - 5 | 337 | 59 | |
| GenBank mRNA | JI871947.1 | - | 4 - 5 | 309 | 59 | |
| JP053162.1 | - | 4 - 5 | 309 | 59 | ||
More Information
Set Membership: |
|

Chmp1a
Genomic Map