Assay Details
| Gene Symbol: Shmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496328.2 | XP_003496376.1 | 6 - 7 | 881 | 58 | |
| GenBank mRNA | JI891972.1 | - | 6 - 7 | 824 | 58 | |
More Information
Set Membership: |
|

Shmt1
Genomic Map