Assay Details
| Gene Symbol: Phyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496411.2 | XP_003496459.1 | 7 - 8 | 894 | 61 | |
| GenBank mRNA | JI890119.1 | - | 7 - 8 | 887 | 61 | |
| JP039947.1 | - | 7 - 8 | 887 | 61 | ||
More Information
Set Membership: |
|

Phyh
Genomic Map