Assay Details
| Gene Symbol: Neurl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498299.2 | XP_003498347.1 | 3 - 4 | 781 | 77 | |
| GenBank mRNA | JI869481.1 | - | 2 - 3 | 79 | 77 | |
| JP054633.1 | - | 2 - 3 | 79 | 77 | ||
More Information
Set Membership: |
|

Neurl3
Genomic Map