Assay Details
| Gene Symbol: Mon1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500660.2 | XP_003500708.1 | 3 - 4 | 1731 | 81 | |
| XM_007641413.1 | XP_007639603.1 | 4 - 5 | 1487 | 81 | ||
| XM_007641414.1 | XP_007639604.1 | 4 - 5 | 1515 | 81 | ||
| GenBank mRNA | JI887040.1 | - | 1 - 2 | 326 | 81 | |
| JP065502.1 | - | 1 - 2 | 326 | 81 | ||
More Information
Set Membership: |
|

Mon1b
Genomic Map